The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178389.3 | NP_848476.1 | 9 - 10 | 1157 | 98 |
GenBank mRNA | AK146184.1 | - | 10 - 11 | 1048 | 98 |
BC027438.1 | - | 9 - 10 | 1157 | 98 |