Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Snapc1
Entrez Gene ID
75627
Gene Name
small nuclear RNA activating complex, polypeptide 1
Gene Aliases
2700033G17Rik, AU015787
Chromosome Location
Chr.12: 73964477 - 73985925 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178392.4 NP_848479.1 9 - 10 1166 77
GenBank mRNA AK012317.1 - 8 - 9 824 77
AK138261.1 - 9 - 10 1167 77
AK152833.1 - 9 - 10 1172 77
AK207384.1 - 2 - 3 233 77
BC030456.1 - 9 - 10 1148 77

View Full Product Details