Product Details
- Assay Reference Genome
Location
Chr.16:38241427 on build GRCm38
- Cytoband
- 16B3
- Species
- Mus musculus
- Variation Type
- Copy Number
Additional Information
Target Gene Details
- Entrez Gene ID
- 56637
- Gene Symbol
- Gsk3b
- Gene Name
- glycogen synthase kinase 3 beta
- Gene Aliases
- 7330414F15Rik, 8430431H08Rik, C86142, GSK-3, GSK-3beta, GSK3
- Location
- Chr.16:38089001-38246080 on build GRCm38
- Assay Gene Location
- Within Exon 11
View Full Product Details