Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Myo1g
Entrez Gene ID
246177
Gene Name
myosin IG
Gene Aliases
E430002D17Rik
Chromosome Location
Chr.11: 6506548 - 6520960 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178440.4 NP_848534.2 17 - 18 2501 83
GenBank mRNA AK030825.1 - 17 - 18 2502 83
AK036461.1 - 17 - 18 2486 83
AK042033.1 - 17 - 18 2453 83
AK088011.1 - 17 - 18 2502 83
AK146251.1 - 17 - 18 2474 83
AK159700.1 - 17 - 18 2477 83
AK171284.1 - 17 - 18 2495 83
AK171505.1 - 17 - 18 2475 83
BC028661.1 - 5 - 6 789 83
BC113755.1 - 17 - 18 2445 83
BC113954.1 - 17 - 18 2445 83

View Full Product Details