The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178601.3 | NP_848716.2 | 2 - 3 | 202 | 64 |
GenBank mRNA | AF334609.1 | - | 1 - 2 | 106 | 64 |
AK004135.1 | - | 1 - 2 | 104 | 64 | |
AK008178.1 | - | 2 - 3 | 165 | 64 | |
AK010504.1 | - | 2 - 3 | 162 | 64 | |
AK013334.1 | - | 2 - 3 | 131 | 64 | |
AK018039.1 | - | 2 - 3 | 160 | 64 | |
AK084933.1 | - | 2 - 3 | 138 | 64 | |
AK151176.1 | - | 2 - 3 | 181 | 64 | |
AK152214.1 | - | 2 - 3 | 181 | 64 | |
AK166903.1 | - | 2 - 3 | 152 | 64 | |
BC037602.1 | - | 1 - 2 | 123 | 64 | |
BC069930.1 | - | 1 - 2 | 112 | 64 | |
DV649342.1 | - | 2 - 3 | 202 | 64 |