Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Reep1
Entrez Gene ID
52250
Gene Name
receptor accessory protein 1
Gene Aliases
D6Ertd253e
Chromosome Location
Chr.6: 71707681 - 71810710 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178608.4 NP_848723.1 4 - 5 596 82
GenBank mRNA AK035238.1 - 4 - 5 430 82
AK045056.1 - 4 - 5 418 82
AK053158.1 - 4 - 5 414 82
AK136916.1 - 4 - 5 375 82
AY562229.1 - 4 - 5 307 82
BC046826.1 - 4 - 5 367 82
CX239705.1 - 4 - 5 596 82

View Full Product Details