Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
E2f7
Entrez Gene ID
52679
Gene Name
E2F transcription factor 7
Gene Aliases
A630014C11Rik, D10Ertd739e
Chromosome Location
Chr.10: 110745427 - 110787384 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178609.4 NP_848724.2 8 - 9 1525 107
GenBank mRNA AK031078.1 - 8 - 9 1523 107
AK045040.1 - 8 - 9 1490 107
AK143686.1 - 8 - 9 1498 107
AY463359.1 - 8 - 9 1522 107
BC145429.1 - 8 - 9 1478 107
BC150772.1 - 8 - 9 1478 107

View Full Product Details