The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178620.3 | NP_848735.1 | 11 - 12 | 1116 | 72 |
GenBank mRNA | AF015191.1 | - | 11 - 12 | 1082 | 72 |
AK031384.1 | - | 11 - 12 | 1311 | 72 | |
AK032043.1 | - | 11 - 12 | 1312 | 72 | |
AK217261.1 | - | 1 - 2 | 153 | 72 | |
BC016209.1 | - | 12 - 13 | 1075 | 72 | |
BC079583.1 | - | 11 - 12 | 1143 | 72 | |
BC088988.1 | - | 11 - 12 | 1116 | 72 | |
BC100547.1 | - | 11 - 12 | 3192 | 72 | |
BC137970.1 | - | 11 - 12 | 1103 | 72 | |
BC137971.1 | - | 11 - 12 | 1104 | 72 | |
BC145068.1 | - | 11 - 12 | 1083 | 72 |