Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Urgcp
Entrez Gene ID
72046
Gene Name
upregulator of cell proliferation
Gene Aliases
2010005J08Rik, 9130001I21Rik, AW060359, Urg4, mKIAA1507
Chromosome Location
Chr.11: 5713417 - 5762436 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077661.1 NP_001071129.1 3 - 4 591 71
NM_178623.3 NP_848738.2 3 - 4 175 71
GenBank mRNA AK033577.1 - 3 - 4 149 71
AK035580.1 - 3 - 4 587 71
AK041953.1 - 4 - 5 840 71
AK191392.1 - 3 - 4 184 71
AK211020.1 - 3 - 4 71
BC066994.1 - 3 - 4 472 71
BY058620.1 - 3 - 4 175 71

View Full Product Details