The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178624.6 | NP_848739.1 | 9 - 10 | 715 | 58 |
GenBank mRNA | AK013142.1 | - | 10 - 11 | 849 | 58 |
AK039010.1 | - | 9 - 10 | 715 | 58 | |
AK044693.1 | - | 9 - 10 | 712 | 58 | |
AK045742.1 | - | 9 - 10 | 702 | 58 | |
AK089994.1 | - | 9 - 10 | 691 | 58 | |
AK141177.1 | - | 9 - 10 | 705 | 58 | |
AK163472.1 | - | 9 - 10 | 711 | 58 | |
BC051530.1 | - | 5 - 6 | 529 | 58 | |
BC096582.1 | - | 9 - 10 | 701 | 58 | |
BC145665.1 | - | 9 - 10 | 695 | 58 | |
BC145998.1 | - | 9 - 10 | 695 | 58 |