Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
B3gntl1
Entrez Gene ID
210004
Gene Name
UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase-like 1
Gene Aliases
6030413G23Rik
Chromosome Location
Chr.11: 121616197 - 121673160 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178664.5 NP_848779.2 5 - 6 513 101
GenBank mRNA AK031361.1 - 5 - 6 502 101
AK043722.1 - 5 - 6 516 101
AK045461.1 - 4 - 5 370 101
AK156318.1 - 5 - 6 491 101
BC088993.1 - 5 - 6 378 101
BC125626.1 - 5 - 6 486 101
BC132193.1 - 5 - 6 486 101
BC145163.1 - 5 - 6 490 101
BC145165.1 - 5 - 6 552 101
BC145166.1 - 5 - 6 486 101

View Full Product Details