The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178683.4 | NP_848798.1 | 2 - 3 | 398 | 79 |
GenBank mRNA | AK036550.1 | - | 2 - 3 | 357 | 79 |
AK043283.1 | - | - | 355 | 79 | |
AK077676.1 | - | 2 - 3 | 399 | 79 | |
AK083480.1 | - | 2 - 3 | 394 | 79 | |
BC152330.1 | - | 2 - 3 | 403 | 79 | |
BC152331.1 | - | 2 - 3 | 403 | 79 |