The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178706.4 | NP_848821.2 | 5 - 6 | 1042 | 57 |
GenBank mRNA | AK032377.1 | - | 4 - 5 | 947 | 57 |
AK045719.1 | - | 6 - 7 | 672 | 57 | |
AK047813.1 | - | 5 - 6 | 967 | 57 | |
BC098229.1 | - | 5 - 6 | 981 | 57 | |
BC120501.1 | - | 5 - 6 | 984 | 57 | |
BC144741.1 | - | 4 - 5 | 705 | 57 | |
BC144742.1 | - | 5 - 6 | 980 | 57 | |
BC144743.1 | - | 5 - 6 | 984 | 57 | |
DQ157764.1 | - | 5 - 6 | 961 | 57 | |
DQ208935.1 | - | 5 - 6 | 987 | 57 |