The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178717.3 | NP_848832.1 | 1 - 1 | 2054 | 76 |
GenBank mRNA | AK043414.1 | - | 1 - 1 | 2079 | 76 |
AK046367.1 | - | 1 - 1 | 1805 | 76 | |
AK084001.1 | - | 1 - 1 | 2056 | 76 | |
AY170825.1 | - | 1 - 1 | 1370 | 76 | |
AY633762.1 | - | 1 - 1 | 1370 | 76 | |
BC053073.1 | - | 1 - 1 | 1476 | 76 |