The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178791.4 | NP_848906.1 | 7 - 8 | 991 | 66 |
GenBank mRNA | AK040699.1 | - | 7 - 8 | 839 | 66 |
AK053685.1 | - | 7 - 8 | 992 | 66 | |
BC107254.2 | - | 7 - 8 | 538 | 66 | |
BC125012.1 | - | 7 - 8 | 537 | 66 | |
BC153828.1 | - | 7 - 8 | 538 | 66 |