The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170855.1 | NP_001164326.1 | 2 - 3 | 534 | 86 |
NM_178872.4 | NP_849203.2 | 2 - 3 | 535 | 86 | |
GenBank mRNA | AB103063.1 | - | 2 - 3 | 329 | 86 |
AY251389.1 | - | 2 - 3 | 292 | 86 | |
BC058106.1 | - | 2 - 3 | 453 | 86 | |
CJ046908.1 | - | 2 - 3 | 533 | 86 |