The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178887.4 | NP_849218.2 | 6 - 7 | 1347 | 62 |
GenBank mRNA | AK046338.1 | - | 6 - 7 | 1309 | 62 |
AK084379.1 | - | 7 - 8 | 1350 | 62 | |
BC060634.1 | - | 6 - 7 | 1347 | 62 |