The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_181040.4 | NP_851383.2 | 9 - 9 | 2149 | 150 |
GenBank mRNA | AK032020.1 | - | - | 1751 | 150 |
BC044874.1 | - | 9 - 9 | 1988 | 150 |