The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_181344.5 | NP_851989.3 | 2 - 3 | 344 | 61 |
GenBank mRNA | AF456428.1 | - | 1 - 2 | 228 | 61 |
AK038659.1 | - | 2 - 3 | 309 | 61 | |
AK133495.1 | - | 2 - 3 | 323 | 61 | |
AK134207.1 | - | 2 - 3 | 345 | 61 | |
AK162497.1 | - | 2 - 3 | 299 | 61 | |
BC070472.1 | - | 2 - 3 | 307 | 61 |