The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_181400.3 | NP_852065.2 | 1 - 2 | 318 | 73 |
GenBank mRNA | AK047357.1 | - | 1 - 2 | 318 | 73 |
AK163422.1 | - | 1 - 2 | 209 | 73 | |
BC040337.1 | - | 1 - 2 | 179 | 73 |