The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286040.1 | NP_001272969.1 | 11 - 12 | 1634 | 98 |
NM_001286041.1 | NP_001272970.1 | 10 - 11 | 1571 | 98 | |
NM_181413.4 | NP_852078.1 | 11 - 12 | 1634 | 98 | |
GenBank mRNA | AK129094.1 | - | 10 - 11 | 1505 | 98 |
AK147273.1 | - | 11 - 12 | 1569 | 98 | |
BC050847.1 | - | 11 - 12 | 1600 | 98 |