Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Anks1
Entrez Gene ID
224650
Gene Name
ankyrin repeat and SAM domain containing 1
Gene Aliases
Anks1a, mKIAA0229
Chromosome Location
Chr.17: 27909306 - 28062779 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286040.1 NP_001272969.1 16 - 17 2647 89
NM_001286041.1 NP_001272970.1 15 - 16 2584 89
NM_181413.4 NP_852078.1 16 - 17 2647 89
GenBank mRNA AK080878.1 - 5 - 6 1282 89
AK129094.1 - 15 - 16 2518 89
AK147273.1 - 16 - 17 2582 89
AK160806.1 - 3 - 4 321 89
AK169828.1 - 5 - 6 593 89
BC010633.1 - 4 - 5 530 89
BC050847.1 - 16 - 17 2613 89
BC057331.1 - 4 - 5 791 89

View Full Product Details