The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_181424.3 | NP_852089.2 | 3 - 4 | 207 | 57 |
GenBank mRNA | AK163448.1 | - | 3 - 4 | 218 | 57 |
BC025144.1 | - | 3 - 4 | 206 | 57 | |
BY726952.1 | - | 3 - 4 | 218 | 57 |