Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fam69c
Entrez Gene ID
240479
Gene Name
family with sequence similarity 69, member C
Gene Aliases
B230399E16Rik
Chromosome Location
Chr.18: 84718987 - 84741731 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173770.4 NP_776131.1 3 - 4 919 97
GenBank mRNA AK046504.1 - 3 - 4 920 97
BC119040.1 - 1 - 2 507 97
BC119042.1 - 1 - 2 507 97

View Full Product Details