The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173773.1 | NP_776134.1 | 4 - 5 | 1089 | 75 |
GenBank mRNA | AK054346.1 | - | 4 - 5 | 1111 | 75 |
AY573564.1 | - | 4 - 5 | 1089 | 75 | |
BC098485.1 | - | 3 - 4 | 948 | 75 |