Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc45a1
Entrez Gene ID
242773
Gene Name
solute carrier family 45, member 1
Gene Aliases
C230078B22, Dnb5
Chromosome Location
Chr.4: 150629396 - 150652174 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173774.3 NP_776135.2 8 - 9 2164 97
GenBank mRNA AK082651.1 - - 2157 97
AK134683.1 - 8 - 9 2155 97
BC051188.1 - 8 - 9 2126 97

View Full Product Details