Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Robo2
Entrez Gene ID
268902
Gene Name
roundabout homolog 2 (Drosophila)
Gene Aliases
2600013A04Rik, 9430089E08Rik, BB097918, D230004I22Rik, mKIAA1568
Chromosome Location
Chr.16: 73892061 - 75447298 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175549.4 NP_780758.3 2 - 3 394 78
GenBank mRNA AK031495.1 - 2 - 3 920 78
AK035110.1 - 2 - 3 1268 78
AK085461.1 - 2 - 3 1019 78
BC055333.1 - 2 - 3 702 78
DQ533875.1 - 3 - 4 674 78
DQ533876.1 - 2 - 3 394 78

View Full Product Details