The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_176942.4 | NP_795916.1 | 5 - 6 | 844 | 75 |
GenBank mRNA | AK038476.1 | - | 5 - 6 | 846 | 75 |
AK083185.1 | - | 5 - 6 | 631 | 75 | |
BC062112.1 | - | 5 - 6 | 842 | 75 |