The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_176954.3 | NP_795928.2 | 3 - 4 | 387 | 76 |
GenBank mRNA | AK083461.1 | - | 2 - 3 | 115 | 76 |
AK132321.1 | - | 3 - 4 | 353 | 76 | |
AK158669.1 | - | 2 - 3 | 164 | 76 | |
BY246044.1 | - | 3 - 4 | 76 |