The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_176972.4 | NP_795946.2 | 16 - 17 | 2044 | 93 |
GenBank mRNA | AK030013.1 | - | 16 - 17 | 2045 | 93 |
AK035640.1 | - | 15 - 16 | 2028 | 93 | |
AK173210.1 | - | 12 - 13 | 1520 | 93 | |
BC114208.1 | - | 14 - 15 | 4159 | 93 | |
BC139091.1 | - | 16 - 17 | 1986 | 93 |