Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cep95
Entrez Gene ID
320162
Gene Name
centrosomal protein 95
Gene Aliases
4732496G21Rik, AI118346, AI448335, Ccdc45, D330027A14Rik, F630025I20Rik
Chromosome Location
Chr.11: 106789252 - 106818861 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166685.1 NP_001160157.1 11 - 12 1418 96
NM_177088.3 NP_796062.2 12 - 13 1547 96
GenBank mRNA AK076400.1 - 12 - 13 1525 96
BC090675.1 - 12 - 13 1503 96
BC116308.1 - 11 - 12 1384 96
BC116309.1 - 11 - 12 1397 96

View Full Product Details