Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cep95
Entrez Gene ID
320162
Gene Name
centrosomal protein 95
Gene Aliases
4732496G21Rik, AI118346, AI448335, Ccdc45, D330027A14Rik, F630025I20Rik
Chromosome Location
Chr.11: 106789252 - 106818861 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166685.1 NP_001160157.1 4 - 5 481 84
NM_177088.3 NP_796062.2 4 - 5 481 84
GenBank mRNA AK052705.1 - 4 - 5 409 84
AK076400.1 - 4 - 5 459 84
AK149412.1 - 1 - 2 963 84
BC090675.1 - 4 - 5 439 84
BC116308.1 - 4 - 5 447 84
BC116309.1 - 4 - 5 447 84

View Full Product Details