Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fndc7
Entrez Gene ID
320181
Gene Name
fibronectin type III domain containing 7
Gene Aliases
E230011A21Rik
Chromosome Location
Chr.3: 108853678 - 108890008 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001165965.1 NP_001159437.1 7 - 8 1670 102
NM_177091.5 NP_796065.2 8 - 9 1928 102
GenBank mRNA AK053989.1 - 8 - 9 1791 102
AK054241.1 - 8 - 9 1928 102
BC111898.1 - 7 - 8 1477 102
BC113188.1 - 8 - 9 1734 102

View Full Product Details