Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Iqcb1
Entrez Gene ID
320299
Gene Name
IQ calmodulin-binding motif containing 1
Gene Aliases
6820449I09Rik, AV128382, NPHP5
Chromosome Location
Chr.16: 36828363 - 36872719 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177128.4 NP_796102.2 5 - 6 566 149
GenBank mRNA AK078554.1 - 5 - 6 559 149
AK165220.1 - 5 - 6 550 149
BC029084.1 - 5 - 6 506 149

View Full Product Details