The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177139.5 | NP_796113.1 | 3 - 4 | 447 | 63 |
NR_033304.2 | 4 - 5 | 612 | 63 | ||
GenBank mRNA | AK053620.1 | - | 4 - 5 | 578 | 63 |
BB619054.1 | - | 3 - 4 | 414 | 63 | |
BB653059.1 | - | 3 - 4 | 408 | 63 | |
BC070462.1 | - | 3 - 4 | 468 | 63 |