The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177151.3 | NP_796125.2 | 3 - 4 | 553 | 118 |
GenBank mRNA | AK049111.1 | - | 4 - 5 | 673 | 118 |
AK053274.1 | - | 3 - 4 | 553 | 118 | |
BC158052.1 | - | 3 - 4 | 533 | 118 |