Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Phf8
Entrez Gene ID
320595
Gene Name
PHD finger protein 8
Gene Aliases
9830141C09Rik, mKIAA1111
Chromosome Location
Chr.X: 151520612 - 151633859 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113354.1 NP_001106825.1 16 - 17 2852 66
NM_177201.5 NP_796175.1 15 - 16 2549 66
GenBank mRNA AK036609.1 - 15 - 16 1923 66
AK122447.1 - 15 - 16 1943 66
AK147939.1 - 16 - 17 2239 66
BC138899.1 - 15 - 16 1818 66

View Full Product Details