The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177310.2 | NP_796284.1 | 3 - 4 | 630 | 74 |
GenBank mRNA | AK088324.1 | - | 3 - 4 | 630 | 74 |
BC137569.1 | - | 3 - 4 | 614 | 74 | |
BC144717.1 | - | 3 - 4 | 615 | 74 |