The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177667.4 | NP_808335.1 | 5 - 6 | 1123 | 94 |
GenBank mRNA | AK028895.1 | - | 5 - 6 | 1124 | 94 |
BC096484.1 | - | 5 - 6 | 1100 | 94 | |
BC132162.1 | - | 5 - 6 | 1064 | 94 |