Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc35d1
Entrez Gene ID
242585
Gene Name
solute carrier family 35 (UDP-glucuronic acid/UDP-N-acetylgalactosamine dual transporter), member D1
Gene Aliases
AI834976, C330011J09, UGTREL7, mKIAA0260
Chromosome Location
Chr.4: 103170649 - 103217826 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177732.4 NP_808400.1 7 - 8 653 69
GenBank mRNA AK049180.1 - 7 - 8 654 69
AK049462.1 - 7 - 8 573 69
AK140305.1 - 7 - 8 945 69
AK169775.1 - 7 - 8 555 69
AK220330.1 - 7 - 8 563 69
BC115984.1 - 8 - 9 554 69

View Full Product Details