The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177766.3 | NP_808434.2 | 2 - 3 | 519 | 84 |
GenBank mRNA | AK031600.1 | - | 2 - 3 | 267 | 84 |
BC115553.1 | - | 2 - 3 | 178 | 84 | |
BC115554.1 | - | 2 - 3 | 178 | 84 |