Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bpifc
Entrez Gene ID
270757
Gene Name
BPI fold containing family C
Gene Aliases
4732454E24, Bpil2
Chromosome Location
Chr.10: 85959340 - 86015898 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177772.4 NP_808440.2 12 - 13 1295 81
GenBank mRNA AK028764.1 - 12 - 13 1292 81
AK132518.1 - 12 - 13 1331 81
BC112376.1 - 12 - 13 1283 81

View Full Product Details