Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rft1
Entrez Gene ID
328370
Gene Name
RFT1 homolog (S. cerevisiae)
Gene Aliases
D930025H04
Chromosome Location
Chr.14: 30654341 - 30691317 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177815.3 NP_808483.2 2 - 3 184 68
GenBank mRNA AK086390.1 - 2 - 3 173 68
BC082539.1 - 2 - 3 152 68
BC094336.1 - 2 - 3 152 68
BC116368.2 - 2 - 3 165 68
BC116369.2 - 2 - 3 165 68
BC141275.1 - 2 - 3 178 68
BC145606.1 - 2 - 3 178 68

View Full Product Details