The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177893.3 | NP_808561.2 | 2 - 3 | 1651 | 124 |
GenBank mRNA | AK077918.1 | - | 2 - 3 | 1642 | 124 |
AK173087.1 | - | 1 - 2 | 1109 | 124 |