The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177906.4 | NP_808574.2 | 3 - 4 | 491 | 71 |
GenBank mRNA | AK028345.1 | - | 2 - 3 | 459 | 71 |
AK158538.1 | - | 3 - 4 | 813 | 71 | |
BC076581.1 | - | 3 - 4 | 491 | 71 |