Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tgm4
Entrez Gene ID
331046
Gene Name
transglutaminase 4 (prostate)
Gene Aliases
9530008N10Rik, Eapa1, TGP
Chromosome Location
Chr.9: 123034741 - 123067558 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177911.4 NP_808579.2 10 - 11 1358 71
GenBank mRNA AF486627.1 - 10 - 11 71
AK035279.1 - 10 - 11 1359 71
BC141297.1 - 10 - 11 1355 71
BC145622.1 - 10 - 11 1355 71

View Full Product Details