The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177920.5 | NP_808588.3 | 4 - 5 | 1124 | 109 |
GenBank mRNA | AK050361.1 | - | 5 - 6 | 669 | 109 |
AK149459.1 | - | 4 - 5 | 1105 | 109 | |
AY575783.1 | - | 3 - 4 | 1058 | 109 | |
BC132100.1 | - | 3 - 4 | 1119 | 109 |