Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Stard9
Entrez Gene ID
668880
Gene Name
START domain containing 9
Gene Aliases
4831403C07Rik, E230025N21Rik, Kif16a, mKIAA1300
Chromosome Location
Chr.2: 120629091 - 120716086 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK029170.1 - - 2096 66
AK029732.1 - 9 - 10 3445 66
AK122489.1 - 9 - 10 4145 66
BC032885.1 - 7 - 8 855 66

View Full Product Details