The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001190376.1 | NP_001177305.1 | 16 - 17 | 3093 | 75 |
NM_001190378.1 | NP_001177307.1 | 16 - 17 | 3093 | 75 | |
NM_001190379.1 | NP_001177308.1 | 15 - 16 | 3021 | 75 | |
NM_178116.4 | NP_835217.1 | 16 - 17 | 3108 | 75 | |
GenBank mRNA | AK122400.1 | - | 14 - 15 | 2749 | 75 |
AK150186.1 | - | 16 - 17 | 2946 | 75 | |
AK169118.1 | - | 15 - 16 | 2860 | 75 | |
BC023962.1 | - | 11 - 12 | 2907 | 75 | |
BC027039.1 | - | 5 - 6 | 817 | 75 | |
BC049133.1 | - | 16 - 17 | 2937 | 75 | |
BC056395.1 | - | 16 - 17 | 2922 | 75 | |
BC059065.1 | - | 15 - 16 | 2888 | 75 |