The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080443.2 | NP_536691.2 | 1 - 2 | 460 | 71 |
NR_003961.1 | 1 - 2 | 460 | 71 | ||
GenBank mRNA | AK051017.1 | - | 1 - 2 | 373 | 71 |
BC085155.1 | - | 1 - 2 | 440 | 71 |